Search
Search
| Gene Symbol: Acaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130433.1 | NP_569117.1 | 1 - 2 | 181 | 67 | |
| GenBank mRNA | FQ216726.1 | - | 1 - 2 | 44 | 67 | |
| X05341.1 | - | 1 - 2 | 181 | 67 | ||
Set Membership: |
|