Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00679785_m1
          See other ACOT12 GE Assays ›
          Gene Symbol:
          Acot12
          Gene Name:
          acyl-CoA thioesterase 12
          Gene Aliases:
          Cach, rACH
          Chromosome Location:
          Chr.2: 20857050 - 20900039 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00679785_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acot12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130747.1 NP_570103.1 11 - 12 1147 70
          XM_008760628.1 XP_008758850.1 12 - 13 1642 70
          GenBank mRNA AB040609.1 - 11 - 12 1147 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000139.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acot12

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          acetyl-CoA metabolic process
          pyruvate metabolic process
          fatty acid metabolic process
          acyl-CoA metabolic process
          acetyl-CoA hydrolase activity
          ATP binding
          lipid binding
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84014 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109617 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13126 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13126 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13126 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13126 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97417 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59260 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812453 - -
          Affymetrix Rat Gene 1.0 ST Array 10812442 - -
          Affymetrix Rat Gene 1.0 ST Array 10812443 - -
          Affymetrix Rat Gene 1.0 ST Array 10812447 - -
          Affymetrix Rat Gene 1.0 ST Array 10812448 - -
          Affymetrix Rat Gene 1.0 ST Array 10812441 - -
          Affymetrix Rat Gene 1.0 ST Array 10812450 - -
          Affymetrix Rat Gene 1.0 ST Array 10812446 - -
          Affymetrix Rat Gene 1.0 ST Array 10812439 - -
          Affymetrix Rat Gene 1.0 ST Array 10812451 - -
          Affymetrix Rat Gene 1.0 ST Array 10812452 - -
          Affymetrix Rat Gene 1.0 ST Array 10812440 - -
          Affymetrix Rat Gene 1.0 ST Array 10812444 - -
          Affymetrix Rat Gene 1.0 ST Array 10812449 - -
          Affymetrix Rat Gene 1.0 ST Array 10812445 - -
          Affymetrix Rat Gene 1.1 ST Array 10812438 - -
          Rat Genome 230 2.0 Array 1369485_PM_at - -
          Rat Genome 230 2.0 Array 1369485_at - -
          Affymetrix HT Rat Focus Array 1369485_at - -
          Affymetrix HT Rat Focus Array 1369485_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200000718.rn.1 TC0200000139.rn.1 RTA TC
          Rat Expression Set 230 1369485_PM_at - -
          Rat Expression Set 230 1369485_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812438 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373584 - -
          RatRef-12 Expression BeadChip ILMN_64941 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline