Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00680598_g1
          See other TAT GE Assays ›
          Gene Symbol:
          Tat
          Gene Name:
          tyrosine aminotransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 41675639 - 41686195 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00680598_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012668.2 NP_036800.1 11 - 12 1266 59
          GenBank mRNA BC089813.1 - 11 - 12 1266 7372826 59
          M18340.1 - 11 - 12 1319 59
          X02741.1 - 11 - 12 1319 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001432.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tat

          Panther Classification:

          Molecular Function -

          transaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          gluconeogenesis
          2-oxoglutarate metabolic process
          cellular amino acid metabolic process
          glutamate metabolic process
          L-phenylalanine catabolic process
          tyrosine catabolic process
          response to oxidative stress
          response to organic cyclic compound
          response to mercury ion
          response to glucocorticoid
          L-tyrosine:2-oxoglutarate aminotransferase activity
          amino acid binding
          pyridoxal phosphate binding
          L-phenylalanine:2-oxoglutarate aminotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P515484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45573 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P515484 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P515484 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P515484 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P515484 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701193605 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34656 - -
          Agilent Rat cDNA Microarray (G4105A) 701193605 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807825 - -
          Affymetrix Rat Gene 1.0 ST Array 10807821 - -
          Affymetrix Rat Gene 1.0 ST Array 10807823 - -
          Affymetrix Rat Gene 1.0 ST Array 10807824 - -
          Affymetrix Rat Gene 1.0 ST Array 10807818 - -
          Affymetrix Rat Gene 1.0 ST Array 10807817 - -
          Affymetrix Rat Gene 1.0 ST Array 10807820 - -
          Affymetrix Rat Gene 1.0 ST Array 10807827 - -
          Affymetrix Rat Gene 1.0 ST Array 10807826 - -
          Affymetrix Rat Gene 1.0 ST Array 10807816 - -
          Affymetrix Rat Gene 1.0 ST Array 10807822 - -
          Affymetrix Rat Gene 1.0 ST Array 10807819 - -
          Affymetrix Rat Gene 1.1 ST Array 10807815 - -
          Rat Genome 230 2.0 Array 1369790_PM_at - -
          Rat Genome 230 2.0 Array 1369790_at - -
          Affymetrix HT Rat Focus Array 1369790_PM_at - -
          Affymetrix HT Rat Focus Array 1369790_at - -
          Affymetrix Rat Clariom S Assay PSR1900011646.rn.1 TC1900001432.rn.1 RTA TC
          Rat Expression Set 230 1369790_PM_at - -
          Rat Expression Set 230 1369790_at - -
          Affymetrix Rat Gene 1.0 ST Array 10807815 - -
          Rat Genome U34 Set X02741_s_at - -
          Rat Genome U34 Set X02741_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21628055 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367659 - -
          RatRef-12 Expression BeadChip ILMN_64587 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline