Search
Search
| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013119.1 | NP_037251.1 | 3 - 4 | 798 | 115 | |
| XM_008761914.1 | XP_008760136.1 | 2 - 3 | 601 | 115 | ||
| XM_008761915.1 | XP_008760137.1 | 3 - 4 | 604 | 115 | ||
| XM_008761916.1 | XP_008760138.1 | 2 - 3 | 602 | 115 | ||
| XM_008761917.1 | XP_008760139.1 | 2 - 3 | 601 | 115 | ||
| XM_008761918.1 | XP_008760140.1 | 2 - 3 | 601 | 115 | ||
| XM_008761919.1 | XP_008760141.1 | 2 - 3 | 601 | 115 | ||
| XM_008761920.1 | XP_008760142.1 | 2 - 3 | 601 | 115 | ||
| GenBank mRNA | Y00766.1 | - | 3 - 4 | 798 | 115 | |
Set Membership: |
|