Search
Search

Exoc2
LOC103689971| Gene Symbol: Exoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134414.2 | NP_599241.2 | 12 - 13 | 1451 | 101 | |
| XM_006253891.1 | XP_006253953.1 | 13 - 14 | 1523 | 101 | ||
| XM_006253893.2 | XP_006253955.1 | 12 - 13 | 1407 | 101 | ||
| XM_006253895.2 | XP_006253957.1 | 11 - 12 | 1335 | 101 | ||
| GenBank mRNA | AF032666.1 | - | 12 - 13 | 1389 | 101 | |
| Gene Symbol: LOC103689971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253876.2 | XP_006253938.1 | 1523 | 101 | ||
| XM_006253878.2 | XP_006253940.1 | 1406 | 101 | |||
| XM_006253880.2 | XP_006253942.1 | 1334 | 101 | |||
| XM_008771607.1 | XP_008769829.1 | 1451 | 101 | |||
Set Membership: |
|