Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00685240_g1
          See other PIGL GE Assays ›
          Gene Symbol:
          Pigl
          Gene Name:
          phosphatidylinositol glycan anchor biosynthesis, class L
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 48774018 - 48831848 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00685240_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pigl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138901.2 NP_620256.1 6 - 7 668 91
          GenBank mRNA BC074020.1 - 6 - 7 668 7104008 91
          D88364.1 - 6 - 7 1049 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000801.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pigl

          Panther Classification:

          Molecular Function -

          deacetylase

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          N-acetylglucosaminylphosphatidylinositol deacetylase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545362 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119336 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24412 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117619 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133842 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067693 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067693 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067693 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P671601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55752 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24962 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10734315 - -
          Affymetrix Rat Gene 1.0 ST Array 10734310 - -
          Affymetrix Rat Gene 1.0 ST Array 10734307 - -
          Affymetrix Rat Gene 1.0 ST Array 10734311 - -
          Affymetrix Rat Gene 1.0 ST Array 10734314 - -
          Affymetrix Rat Gene 1.0 ST Array 10734308 - -
          Affymetrix Rat Gene 1.0 ST Array 10734312 - -
          Affymetrix Rat Gene 1.0 ST Array 10734309 - -
          Affymetrix Rat Gene 1.0 ST Array 10734313 - -
          Affymetrix Rat Gene 1.1 ST Array 10734306 - -
          Rat Genome 230 2.0 Array 1398309_at - -
          Rat Genome 230 2.0 Array 1398309_PM_at - -
          Affymetrix HT Rat Focus Array 1398309_PM_at - -
          Affymetrix HT Rat Focus Array 1398309_at - -
          Affymetrix Rat Clariom S Assay PSR1000006759.rn.1 TC1000000801.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006762.rn.1 TC1000000801.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006760.rn.1 TC1000000801.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006761.rn.1 TC1000000801.rn.1 RTA TC
          Rat Expression Set 230 1398309_at - -
          Rat Expression Set 230 1398309_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10734306 - -
          Rat Genome U34 Set D88364_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349061 - -
          RatRef-12 Expression BeadChip ILMN_53447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline