Search
Search
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172320.1 | NP_758823.1 | 8 - 9 | 1055 | 82 | |
| XM_008770001.1 | XP_008768223.1 | 8 - 9 | 1103 | 82 | ||
| GenBank mRNA | BC091391.1 | - | 5 - 6 | 620 | 82 | |
| FQ213773.1 | - | 4 - 5 | 491 | 82 | ||
| FQ219466.1 | - | 5 - 6 | 589 | 82 | ||
Set Membership: |
|