Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00688087_m1
          See other APC GE Assays ›
          Gene Symbol:
          Apc
          Gene Name:
          adenomatous polyposis coli
          Gene Aliases:
          RATAPC
          Chromosome Location:
          Chr.18: 27011706 - 27106323 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00688087_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012499.1 NP_036631.1 5 - 6 697 78
          XM_006254506.2 XP_006254568.1 5 - 6 903 78
          XM_006254508.2 XP_006254570.1 6 - 7 744 78
          XM_006254509.2 XP_006254571.1 6 - 7 713 78
          XM_006254510.2 XP_006254572.1 5 - 6 903 78
          XM_006254511.2 XP_006254573.1 6 - 7 744 78
          XM_006254512.2 XP_006254574.1 5 - 6 1352 78
          XM_006254513.2 XP_006254575.1 5 - 6 903 78
          XM_006254515.2 XP_006254577.1 6 - 7 569 78
          XM_006254518.2 XP_006254580.1 5 - 6 1167 78
          XM_006254520.2 XP_006254582.1 5 - 6 903 78
          XM_006254521.2 XP_006254583.1 6 - 7 744 78
          XM_008772015.1 XP_008770237.1 5 - 6 2835 78
          XM_008772016.1 XP_008770238.1 5 - 6 2785 78
          XM_008772017.1 XP_008770239.1 5 - 6 1176 78
          GenBank mRNA D38629.1 - 5 - 6 697 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000255.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apc

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          kidney development
          hair follicle development
          protein complex assembly
          cellular response to DNA damage stimulus
          negative regulation of microtubule depolymerization
          cell cycle arrest
          metaphase/anaphase transition of mitotic cell cycle
          mitotic spindle assembly checkpoint
          establishment or maintenance of cell polarity
          pattern specification process
          axonogenesis
          negative regulation of cell proliferation
          axis specification
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          Wnt signaling pathway
          cell migration
          stem cell population maintenance
          negative regulation of Wnt signaling pathway
          regulation of cell migration
          positive regulation of cell migration
          regulation of epithelial cell differentiation
          positive regulation of epithelial cell differentiation
          pancreas development
          positive regulation of microtubule polymerization
          cytoplasmic microtubule organization
          neuron projection development
          positive regulation of pseudopodium assembly
          regulation of microtubule-based process
          T cell differentiation in thymus
          somatic stem cell population maintenance
          negative regulation of odontogenesis
          response to drug
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of MAPK cascade
          skin development
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          canonical Wnt signaling pathway involved in positive regulation of apoptotic process
          regulation of cell differentiation
          positive regulation of cell differentiation
          regulation of osteoblast differentiation
          regulation of osteoclast differentiation
          positive regulation of protein catabolic process
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cell adhesion
          muscle cell cellular homeostasis
          thymus development
          negative regulation of epithelial cell proliferation
          regulation of nitrogen compound metabolic process
          chromosome organization
          regulation of cell cycle
          positive regulation of cell division
          regulation of attachment of spindle microtubules to kinetochore
          retina development in camera-type eye
          canonical Wnt signaling pathway
          negative regulation of epithelial cell proliferation involved in prostate gland development
          negative regulation of canonical Wnt signaling pathway
          response to alcohol
          cellular response to nerve growth factor stimulus
          regulation of glutamate metabolic process
          protease binding
          protein binding
          beta-catenin binding
          microtubule binding
          protein kinase regulator activity
          protein kinase binding
          protein complex binding
          gamma-catenin binding
          cadherin binding
          microtubule plus-end binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1800001845.rn.1 TC1800000255.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline