Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00688575_m1
          See other PRPS2 GE Assays ›
          Gene Symbol:
          Prps2
          Gene Name:
          phosphoribosyl pyrophosphate synthetase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 28435275 - 28472882 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00688575_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prps2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012634.1 NP_036766.1 5 - 6 1047 60
          XM_006256829.2 XP_006256891.1 5 - 6 819 60
          GenBank mRNA M17259.1 - 5 - 6 788 60
          X16555.1 - 5 - 6 1047 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000277.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prps2

          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          5-phosphoribose 1-diphosphate biosynthetic process
          AMP biosynthetic process
          nucleotide biosynthetic process
          phosphorylation
          ribose phosphate metabolic process
          organ regeneration
          magnesium ion binding
          ribose phosphate diphosphokinase activity
          protein binding
          ATP binding
          AMP binding
          kinase activity
          GDP binding
          carbohydrate binding
          protein homodimerization activity
          ADP binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944917 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61348 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31821 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11495 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11495 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11495 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700886091 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78400 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66776 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62461 - -
          Agilent Rat cDNA Microarray (G4105A) 700886091 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933336 - -
          Affymetrix Rat Gene 1.0 ST Array 10933334 - -
          Affymetrix Rat Gene 1.0 ST Array 10933338 - -
          Affymetrix Rat Gene 1.0 ST Array 10933340 - -
          Affymetrix Rat Gene 1.0 ST Array 10933335 - -
          Affymetrix Rat Gene 1.0 ST Array 10933342 - -
          Affymetrix Rat Gene 1.0 ST Array 10933341 - -
          Affymetrix Rat Gene 1.0 ST Array 10933339 - -
          Affymetrix Rat Gene 1.0 ST Array 10933337 - -
          Affymetrix Rat Gene 1.1 ST Array 10933333 - -
          Rat Genome 230 2.0 Array 1398262_PM_at - -
          Rat Genome 230 2.0 Array 1398262_at - -
          Rat Genome 230 2.0 Array 1368551_PM_at - -
          Rat Genome 230 2.0 Array 1368551_at - -
          Rat Genome 230 2.0 Array 1375932_PM_at - -
          Rat Genome 230 2.0 Array 1375932_at - -
          Affymetrix HT Rat Focus Array 1398262_PM_at - -
          Affymetrix HT Rat Focus Array 1368551_at - -
          Affymetrix HT Rat Focus Array 1398262_at - -
          Affymetrix HT Rat Focus Array 1368551_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00002117.rn.1 TC0X00000277.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002120.rn.1 TC0X00000277.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002116.rn.1 TC0X00000277.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002118.rn.1 TC0X00000277.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00002119.rn.1 TC0X00000277.rn.1 RTA TC
          Rat Expression Set 230 1368551_at - -
          Rat Expression Set 230 1368551_PM_at - -
          Rat Expression Set 230 1398262_PM_at - -
          Rat Expression Set 230 1398262_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933333 - -
          Rat Genome U34 Set X16555_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358229 - -
          RatRef-12 Expression BeadChip ILMN_56524 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline