Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00688966_m1
          See other TGFBR1 GE Assays ›
          Gene Symbol:
          Tgfbr1
          Gene Name:
          transforming growth factor, beta receptor 1
          Gene Aliases:
          Alk5, Skr4, Tgfr-1, tbetaR-I
          Chromosome Location:
          Chr.5: 63056079 - 63124107 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00688966_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfbr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012775.2 NP_036907.2 1449 149
          XM_006238030.2 XP_006238092.2 1463 149
          GenBank mRNA BC087035.1 - 1391 7098473 149
          L26110.1 - 1418 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000606.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgfbr1

          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          skeletal system development
          angiogenesis
          response to hypoxia
          in utero embryonic development
          kidney development
          blastocyst development
          epithelial to mesenchymal transition
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lens development in camera-type eye
          regulation of transcription, DNA-templated
          protein phosphorylation
          apoptotic process
          signal transduction
          transmembrane receptor protein serine/threonine kinase signaling pathway
          transforming growth factor beta receptor signaling pathway
          heart development
          embryo implantation
          aging
          positive regulation of cell proliferation
          germ cell migration
          male gonad development
          response to toxic substance
          post-embryonic development
          anterior/posterior pattern specification
          regulation of gene expression
          positive regulation of gene expression
          regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          response to organic cyclic compound
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          signal transduction by protein phosphorylation
          collagen fibril organization
          positive regulation of cell growth
          lung development
          organ regeneration
          regulation of protein ubiquitination
          negative regulation of chondrocyte differentiation
          activin receptor signaling pathway
          response to prostaglandin E
          intracellular signal transduction
          regulation of growth
          endothelial cell activation
          negative regulation of apoptotic process
          regulation of protein binding
          endothelial cell migration
          response to estrogen
          negative regulation of endothelial cell differentiation
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          thymus development
          digestive tract development
          neuron fate commitment
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          mesenchymal cell differentiation
          artery morphogenesis
          cell motility
          positive regulation of cellular component movement
          positive regulation of filopodium assembly
          response to electrical stimulus
          positive regulation of protein kinase B signaling
          parathyroid gland development
          palate development
          pharyngeal system development
          cardiac epithelial to mesenchymal transition
          pathway-restricted SMAD protein phosphorylation
          positive regulation of SMAD protein import into nucleus
          response to cholesterol
          cellular response to transforming growth factor beta stimulus
          positive regulation of apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type I
          receptor signaling protein activity
          type II transforming growth factor beta receptor binding
          ATP binding
          growth factor binding
          ubiquitin protein ligase binding
          protein complex binding
          SMAD binding
          metal ion binding
          protein heterodimerization activity
          transforming growth factor beta binding
          I-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P045651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113368 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99896 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5312 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045651 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P045651 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P045651 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11556 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P640828 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P703479 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 630 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41519 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81559 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75258 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41085 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868933 - -
          Affymetrix Rat Gene 1.0 ST Array 10868927 - -
          Affymetrix Rat Gene 1.0 ST Array 10868934 - -
          Affymetrix Rat Gene 1.0 ST Array 10868932 - -
          Affymetrix Rat Gene 1.0 ST Array 10868925 - -
          Affymetrix Rat Gene 1.0 ST Array 10868928 - -
          Affymetrix Rat Gene 1.0 ST Array 10868926 - -
          Affymetrix Rat Gene 1.0 ST Array 10868930 - -
          Affymetrix Rat Gene 1.0 ST Array 10868931 - -
          Affymetrix Rat Gene 1.0 ST Array 10868929 - -
          Affymetrix Rat Gene 1.0 ST Array 10868924 - -
          Affymetrix Rat Gene 1.1 ST Array 10868923 - -
          Rat Genome 230 2.0 Array 1376636_at - -
          Rat Genome 230 2.0 Array 1376636_PM_at - -
          Rat Genome 230 2.0 Array 1369504_PM_at - -
          Rat Genome 230 2.0 Array 1369504_at - -
          Affymetrix HT Rat Focus Array 1369504_PM_at - -
          Affymetrix HT Rat Focus Array 1376636_at - -
          Affymetrix HT Rat Focus Array 1369504_at - -
          Affymetrix HT Rat Focus Array 1376636_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500004104.rn.1 TC0500000606.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500004105.rn.1 TC0500000606.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500004106.rn.1 TC0500000606.rn.1 RTA TC
          Rat Expression Set 230 1369504_at - -
          Rat Expression Set 230 1369504_PM_at - -
          Rat Expression Set 230 1376636_PM_at - -
          Rat Expression Set 230 1376636_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868923 - -
          Rat Genome U34 Set S81584_s_at - -
          Rat Genome U34 Set L26110_at - -
          Rat Genome U34 Set rc_AI236800_at - -
          Rat Genome U34 Set S81584_s_at - -
          Rat Genome U34 Set L26110_at - -
          Rat Genome U34 Set L26110_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21323429 - -
          ABI Rat Genome Survey Microarray 22297523 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039256 - -
          RatRef-12 Expression BeadChip ILMN_65380 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline