Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00689323_g1
          See other SLC2A2 GE Assays ›
          Gene Symbol:
          Slc2a2
          Gene Name:
          solute carrier family 2 (facilitated glucose transporter), member 2
          Gene Aliases:
          GTT2, Glut2
          Chromosome Location:
          Chr.2: 114413431 - 114445418 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00689323_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc2a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012879.2 NP_037011.2 10 - 11 1403 74
          XM_006232207.2 XP_006232269.1 11 - 12 1707 74
          GenBank mRNA BC078875.1 - 10 - 11 1403 7127751 74
          J03145.1 - 10 - 11 1478 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000905.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc2a2

          Gene Ontology Categories:

          Function(s) Process(es)

          response to glucose
          carbohydrate utilization
          fructose transport
          glucose transport
          hexose transmembrane transport
          response to peptide hormone
          transmembrane transport
          dehydroascorbic acid transport
          insulin receptor binding
          glucose transmembrane transporter activity
          dehydroascorbic acid transporter activity
          D-glucose transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18334 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011959 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011959 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011959 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P226612 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P488355 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10814635 - -
          Affymetrix Rat Gene 1.0 ST Array 10814634 - -
          Affymetrix Rat Gene 1.0 ST Array 10814638 - -
          Affymetrix Rat Gene 1.0 ST Array 10814641 - -
          Affymetrix Rat Gene 1.0 ST Array 10814636 - -
          Affymetrix Rat Gene 1.0 ST Array 10814642 - -
          Affymetrix Rat Gene 1.0 ST Array 10814643 - -
          Affymetrix Rat Gene 1.0 ST Array 10814631 - -
          Affymetrix Rat Gene 1.0 ST Array 10814639 - -
          Affymetrix Rat Gene 1.0 ST Array 10814632 - -
          Affymetrix Rat Gene 1.0 ST Array 10814633 - -
          Affymetrix Rat Gene 1.0 ST Array 10814637 - -
          Affymetrix Rat Gene 1.0 ST Array 10814640 - -
          Affymetrix Rat Gene 1.1 ST Array 10814630 - -
          Rat Genome 230 2.0 Array 1387228_PM_at - -
          Rat Genome 230 2.0 Array 1387228_at - -
          Affymetrix HT Rat Focus Array 1387228_at - -
          Affymetrix HT Rat Focus Array 1387228_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200005527.rn.1 TC0200000905.rn.1 RTA TC
          Rat Expression Set 230 1387228_PM_at - -
          Rat Expression Set 230 1387228_at - -
          Affymetrix Rat Gene 1.0 ST Array 10814630 - -
          Rat Genome U34 Set L28135_at - -
          Rat Genome U34 Set L28126exon#1_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20931266 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363292 - -
          RatRef-12 Expression BeadChip ILMN_56822 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline