Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00689568_g1
          See other NRD1 GE Assays ›
          Gene Symbol:
          Nrd1
          Gene Name:
          nardilysin 1
          Gene Aliases:
          NRDC
          Chromosome Location:
          Chr.5: 128629952 - 128694209 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00689568_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012993.2 NP_037125.1 30 - 31 3354 74
          XM_006238499.2 XP_006238561.1 32 - 33 3692 74
          XM_006238500.2 XP_006238562.1 31 - 32 3556 74
          GenBank mRNA FQ218095.1 - 30 - 31 3477 74
          L27124.1 - 30 - 31 3355 74
          X93208.1 - 32 - 33 3559 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001179.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrd1

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          positive regulation of membrane protein ectodomain proteolysis
          regulation of endopeptidase activity
          metalloendopeptidase activity
          peptidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115471 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42871 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092374 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092374 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092374 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P200233 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700492806 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51850 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61211 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36394 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63414 - -
          Agilent Rat cDNA Microarray (G4105A) 700492806 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10870868 - -
          Affymetrix Rat Gene 1.0 ST Array 10870879 - -
          Affymetrix Rat Gene 1.0 ST Array 10870876 - -
          Affymetrix Rat Gene 1.0 ST Array 10870865 - -
          Affymetrix Rat Gene 1.0 ST Array 10870875 - -
          Affymetrix Rat Gene 1.0 ST Array 10870858 - -
          Affymetrix Rat Gene 1.0 ST Array 10870877 - -
          Affymetrix Rat Gene 1.0 ST Array 10870882 - -
          Affymetrix Rat Gene 1.0 ST Array 10870878 - -
          Affymetrix Rat Gene 1.0 ST Array 10870862 - -
          Affymetrix Rat Gene 1.0 ST Array 10870881 - -
          Affymetrix Rat Gene 1.0 ST Array 10870872 - -
          Affymetrix Rat Gene 1.0 ST Array 10870873 - -
          Affymetrix Rat Gene 1.0 ST Array 10870884 - -
          Affymetrix Rat Gene 1.0 ST Array 10870888 - -
          Affymetrix Rat Gene 1.0 ST Array 10870886 - -
          Affymetrix Rat Gene 1.0 ST Array 10870861 - -
          Affymetrix Rat Gene 1.0 ST Array 10870885 - -
          Affymetrix Rat Gene 1.0 ST Array 10870864 - -
          Affymetrix Rat Gene 1.0 ST Array 10870857 - -
          Affymetrix Rat Gene 1.0 ST Array 10870870 - -
          Affymetrix Rat Gene 1.0 ST Array 10870871 - -
          Affymetrix Rat Gene 1.0 ST Array 10870889 - -
          Affymetrix Rat Gene 1.0 ST Array 10870880 - -
          Affymetrix Rat Gene 1.0 ST Array 10870883 - -
          Affymetrix Rat Gene 1.0 ST Array 10870887 - -
          Affymetrix Rat Gene 1.0 ST Array 10870874 - -
          Affymetrix Rat Gene 1.0 ST Array 10870869 - -
          Affymetrix Rat Gene 1.0 ST Array 10870867 - -
          Affymetrix Rat Gene 1.0 ST Array 10870863 - -
          Affymetrix Rat Gene 1.0 ST Array 10870866 - -
          Affymetrix Rat Gene 1.1 ST Array 10870856 - -
          Rat Genome 230 2.0 Array 1387110_PM_at - -
          Rat Genome 230 2.0 Array 1387110_at - -
          Affymetrix HT Rat Focus Array 1387110_at - -
          Affymetrix HT Rat Focus Array 1387110_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500007999.rn.1 TC0500001179.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500008001.rn.1 TC0500001179.rn.1 RTA TC
          Rat Expression Set 230 1387110_at - -
          Rat Expression Set 230 1387110_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870856 - -
          Rat Genome U34 Set L27124_s_at - -
          Rat Genome U34 Set rc_AA945902_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20714289 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360625 - -
          RatRef-12 Expression BeadChip ILMN_50219 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline