Search
Search
| Gene Symbol: Scn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271045.1 | NP_001257974.1 | 2 - 3 | 342 | 59 | |
| NM_001271046.2 | NP_001257975.1 | 2 - 3 | 209 | 59 | ||
| NM_017288.2 | NP_058984.1 | 2 - 3 | 423 | 59 | ||
| GenBank mRNA | AF182949.1 | - | 2 - 3 | 203 | 59 | |
| BC094523.1 | - | 2 - 3 | 342 | 7316686 | 59 | |
| FQ216374.1 | - | 2 - 3 | 209 | 59 | ||
| M91808.1 | - | 2 - 3 | 422 | 59 | ||
Set Membership: |
|