Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00692975_m1
          See other TSX GE Assays ›
          Gene Symbol:
          Tsx
          Gene Name:
          testis specific X-linked gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 74269513 - 74283819 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00692975_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019203.1 NP_062076.1 6 - 7 479 82
          XM_006257142.2 XP_006257204.1 7 - 8 559 82
          GenBank mRNA X99797.1 - 6 - 7 479 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000678.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsx

          Gene Ontology Categories:

          Function(s)

          male gonad development
          fertilization
          adult behavior

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11975 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11975 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17027 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58338 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934512 - -
          Affymetrix Rat Gene 1.0 ST Array 10934515 - -
          Affymetrix Rat Gene 1.0 ST Array 10934513 - -
          Affymetrix Rat Gene 1.0 ST Array 10934510 - -
          Affymetrix Rat Gene 1.0 ST Array 10934511 - -
          Affymetrix Rat Gene 1.0 ST Array 10934516 - -
          Affymetrix Rat Gene 1.0 ST Array 10934514 - -
          Affymetrix Rat Gene 1.1 ST Array 10934509 - -
          Rat Genome 230 2.0 Array 1368736_at - -
          Rat Genome 230 2.0 Array 1368736_PM_at - -
          Affymetrix HT Rat Focus Array 1368736_at - -
          Affymetrix HT Rat Focus Array 1368736_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00004359.rn.1 TC0X00000678.rn.1 RTA TC
          Rat Expression Set 230 1368736_at - -
          Rat Expression Set 230 1368736_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10934509 - -
          Rat Genome U34 Set X99797_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20811845 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368901 - -
          RatRef-12 Expression BeadChip ILMN_60573 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline