Search
Search
| Gene Symbol: Espn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239479.2 | XP_006239541.1 | 7 - 8 | 1637 | 113 | |
| XM_008764377.1 | XP_008762599.1 | 7 - 8 | 1646 | 113 | ||
| XM_008764378.1 | XP_008762600.1 | 7 - 8 | 1646 | 113 | ||
| XM_008764380.1 | XP_008762602.1 | 7 - 8 | 1646 | 113 | ||
| GenBank mRNA | AF540946.1 | - | 2 - 3 | 466 | 113 | |
| AF540947.1 | - | 2 - 3 | 466 | 113 | ||
| U46007.1 | - | 7 - 8 | 1460 | 113 | ||
Set Membership: |
|