Search
Search
| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017208.2 | NP_058904.2 | 11 - 12 | 1239 | 59 | |
| GenBank mRNA | BC101906.1 | - | 11 - 12 | 1239 | 7933709 | 59 |
| FQ219857.1 | - | 4 - 5 | 388 | 59 | ||
Set Membership: |
|