Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710227_g1
          See other MAPKAPK2 GE Assays ›
          Gene Symbol:
          Mapkapk2
          Gene Name:
          mitogen-activated protein kinase-activated protein kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 47785161 - 47871684 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710227_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapkapk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178102.2 NP_835203.1 10 - 11 1314 60
          GenBank mRNA AY197741.1 - 10 - 11 1314 60
          BC062048.1 - 10 - 11 1280 5598069 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001374.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mapkapk2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          toll-like receptor signaling pathway
          protein phosphorylation
          inflammatory response
          cellular response to DNA damage stimulus
          peptidyl-serine phosphorylation
          response to lipopolysaccharide
          regulation of interleukin-6 production
          regulation of tumor necrosis factor production
          response to cytokine
          cellular response to vascular endothelial growth factor stimulus
          macropinocytosis
          vascular endothelial growth factor receptor signaling pathway
          mRNA stabilization
          inner ear development
          3'-UTR-mediated mRNA stabilization
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P647072 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107793 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33031 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093367 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P647072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P647072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093367 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093367 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P647072 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P647072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100353 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5348 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32968 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53895 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767492 - -
          Affymetrix Rat Gene 1.0 ST Array 10767500 - -
          Affymetrix Rat Gene 1.0 ST Array 10767498 - -
          Affymetrix Rat Gene 1.0 ST Array 10767497 - -
          Affymetrix Rat Gene 1.0 ST Array 10767496 - -
          Affymetrix Rat Gene 1.0 ST Array 10767494 - -
          Affymetrix Rat Gene 1.0 ST Array 10767493 - -
          Affymetrix Rat Gene 1.0 ST Array 10767501 - -
          Affymetrix Rat Gene 1.0 ST Array 10767490 - -
          Affymetrix Rat Gene 1.0 ST Array 10767495 - -
          Affymetrix Rat Gene 1.0 ST Array 10767491 - -
          Affymetrix Rat Gene 1.0 ST Array 10767499 - -
          Affymetrix Rat Gene 1.1 ST Array 10767489 - -
          Rat Genome 230 2.0 Array 1371446_PM_at - -
          Rat Genome 230 2.0 Array 1371446_at - -
          Affymetrix HT Rat Focus Array 1371446_PM_at - -
          Affymetrix HT Rat Focus Array 1371446_at - -
          Affymetrix Rat Clariom S Assay PSR1300009081.rn.1 TC1300001374.rn.1 RTA TC
          Rat Expression Set 230 1371446_at - -
          Rat Expression Set 230 1371446_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767489 - -
          Rat Genome U34 Set rc_AA799814_at - -
          Rat Genome U34 Set rc_AI237813_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21397986 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline