Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710549_m1
          See other SPATS1 GE Assays ›
          Gene Symbol:
          Spats1
          Gene Name:
          spermatogenesis associated, serine-rich 1
          Gene Aliases:
          Srsp1
          Chromosome Location:
          Chr.9: 17911950 - 17947101 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710549_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spats1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181376.2 NP_852041.1 2 - 3 233 145
          GenBank mRNA AY157978.1 - 2 - 3 233 145

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000255.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spats1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P283176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97454 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P283176 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P283176 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P283176 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P283176 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21885 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12395 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54147 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10921871 - -
          Affymetrix Rat Gene 1.0 ST Array 10921867 - -
          Affymetrix Rat Gene 1.0 ST Array 10921868 - -
          Affymetrix Rat Gene 1.0 ST Array 10921869 - -
          Affymetrix Rat Gene 1.0 ST Array 10921870 - -
          Affymetrix Rat Gene 1.0 ST Array 10921872 - -
          Affymetrix Rat Gene 1.0 ST Array 10921876 - -
          Affymetrix Rat Gene 1.0 ST Array 10921877 - -
          Affymetrix Rat Gene 1.0 ST Array 10921873 - -
          Affymetrix Rat Gene 1.1 ST Array 10921864 - -
          Affymetrix Rat Clariom S Assay PSR0900002391.rn.1 TC0900000255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900002385.rn.1 TC0900000255.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900002392.rn.1 TC0900000255.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10921864 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21396836 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364497 - -
          RatRef-12 Expression BeadChip ILMN_67631 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline