Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hbg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172093.1 | NP_742090.1 | 2 - 3 | 313 | 92 | |
GenBank mRNA | BC157810.1 | - | 2 - 3 | 347 | 9027502 | 92 |
FQ221361.1 | - | 2 - 3 | 365 | 92 | ||
FQ222999.1 | - | 2 - 3 | 360 | 92 | ||
FQ228385.1 | - | 2 - 3 | 367 | 92 | ||
FQ228657.1 | - | 2 - 3 | 363 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |