Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820640_g1
          See other FTH1 GE Assays ›
          Gene Symbol:
          Fth1
          Gene Name:
          ferritin, heavy polypeptide 1
          Gene Aliases:
          Fth
          Chromosome Location:
          Chr.1: 226030940 - 226033228 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820640_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fth1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012848.2 NP_036980.1 1 - 2 241 95
          GenBank mRNA BC060581.1 - 284 95
          BC078892.1 - 1 - 2 257 95
          BC081845.1 - 1 - 2 230 7191119 95
          BC089817.1 - 1 - 2 271 95
          BC097341.1 - 1 - 2 271 95
          BC111078.1 - 1 - 2 265 95
          BC127507.1 - 1 - 2 250 95
          FQ209506.1 - 1 - 2 281 95
          FQ210210.1 - 1 - 2 280 95
          FQ210496.1 - 1 - 2 280 95
          FQ210859.1 - 1 - 2 280 95
          FQ213698.1 - 1 - 2 280 95
          FQ216698.1 - 1 - 2 280 95
          FQ216714.1 - 1 - 2 309 95
          FQ217105.1 - 1 - 2 280 95
          FQ217377.1 - 1 - 2 280 95
          FQ218113.1 - 1 - 2 280 95
          FQ218205.1 - 1 - 2 280 95
          FQ218247.1 - 1 - 2 278 95
          FQ218258.1 - 1 - 2 280 95
          FQ219782.1 - 1 - 2 280 95
          FQ219802.1 - 1 - 2 280 95
          FQ219887.1 - 1 - 2 281 95
          FQ219905.1 - 1 - 2 280 95
          FQ219911.1 - 1 - 2 280 95
          FQ220036.1 - 1 - 2 280 95
          FQ220121.1 - 1 - 2 280 95
          FQ220190.1 - 1 - 2 280 95
          FQ220290.1 - 1 - 2 280 95
          FQ220318.1 - 1 - 2 280 95
          FQ220323.1 - 1 - 2 280 95
          FQ220336.1 - 1 - 2 280 95
          FQ220343.1 - 1 - 2 280 95
          FQ220429.1 - 1 - 2 280 95
          FQ220465.1 - 1 - 2 280 95
          FQ220473.1 - 1 - 2 280 95
          FQ220538.1 - 1 - 2 280 95
          FQ220612.1 - 1 - 2 280 95
          FQ220649.1 - 1 - 2 280 95
          FQ220657.1 - 1 - 2 280 95
          FQ220664.1 - 1 - 2 280 95
          FQ220737.1 - 1 - 2 280 95
          FQ220761.1 - 1 - 2 280 95
          FQ220768.1 - 1 - 2 280 95
          FQ220780.1 - 1 - 2 280 95
          FQ220794.1 - 1 - 2 280 95
          FQ220847.1 - 1 - 2 280 95
          FQ220848.1 - 1 - 2 280 95
          FQ220860.1 - 1 - 2 280 95
          FQ220894.1 - 1 - 2 280 95
          FQ220937.1 - 1 - 2 280 95
          FQ220949.1 - 1 - 2 280 95
          FQ220974.1 - 1 - 2 280 95
          FQ220980.1 - 1 - 2 280 95
          FQ221007.1 - 1 - 2 280 95
          FQ221008.1 - 1 - 2 280 95
          FQ221025.1 - 1 - 2 280 95
          FQ221042.1 - 1 - 2 280 95
          FQ221055.1 - 1 - 2 280 95
          FQ221063.1 - 1 - 2 280 95
          FQ221270.1 - 1 - 2 280 95
          FQ221273.1 - 1 - 2 280 95
          FQ221283.1 - 1 - 2 278 95
          FQ221322.1 - 1 - 2 280 95
          FQ221323.1 - 1 - 2 281 95
          FQ221449.1 - 1 - 2 138 95
          FQ221530.1 - 1 - 2 280 95
          FQ221594.1 - 1 - 2 280 95
          FQ221786.1 - 1 - 2 280 95
          FQ221796.1 - 1 - 2 280 95
          FQ221857.1 - 1 - 2 280 95
          FQ221858.1 - 1 - 2 280 95
          FQ221997.1 - 1 - 2 280 95
          FQ222029.1 - 1 - 2 280 95
          FQ222053.1 - 1 - 2 280 95
          FQ222126.1 - 1 - 2 280 95
          FQ222138.1 - 1 - 2 280 95
          FQ222301.1 - 1 - 2 280 95
          FQ222380.1 - 1 - 2 280 95
          FQ222409.1 - 1 - 2 280 95
          FQ222432.1 - 1 - 2 280 95
          FQ222473.1 - 1 - 2 280 95
          FQ222548.1 - 1 - 2 280 95
          FQ222554.1 - 1 - 2 280 95
          FQ222812.1 - 1 - 2 280 95
          FQ222902.1 - 1 - 2 280 95
          FQ223030.1 - 1 - 2 279 95
          FQ223288.1 - 1 - 2 280 95
          FQ223358.1 - 1 - 2 280 95
          FQ223447.1 - 1 - 2 280 95
          FQ224061.1 - 1 - 2 280 95
          FQ225113.1 - 1 - 2 274 95
          FQ225349.1 - 1 - 2 280 95
          FQ225367.1 - 1 - 2 280 95
          FQ225566.1 - 1 - 2 280 95
          FQ227131.1 - 1 - 2 280 95
          FQ227349.1 - 1 - 2 280 95
          FQ227356.1 - 1 - 2 280 95
          FQ227471.1 - 1 - 2 280 95
          FQ227537.1 - 1 - 2 280 95
          FQ227952.1 - 1 - 2 283 95
          FQ228079.1 - 1 - 2 280 95
          FQ228408.1 - 1 - 2 280 95
          FQ228428.1 - 1 - 2 279 95
          FQ228441.1 - 1 - 2 280 96
          FQ228455.1 - 1 - 2 280 95
          FQ228509.1 - 1 - 2 280 95
          FQ228648.1 - 1 - 2 280 95
          FQ228988.1 - 1 - 2 280 95
          FQ229043.1 - 1 - 2 280 95
          FQ229051.1 - 1 - 2 280 95
          FQ229136.1 - 1 - 2 280 95
          FQ229148.1 - 1 - 2 280 95
          FQ229177.1 - 1 - 2 280 95
          FQ229233.1 - 1 - 2 281 95
          FQ229274.1 - 1 - 2 280 95
          FQ229334.1 - 1 - 2 280 95
          FQ229340.1 - 1 - 2 280 95
          FQ229343.1 - 1 - 2 280 95
          FQ229366.1 - 1 - 2 280 95
          FQ229418.1 - 1 - 2 280 95
          FQ229423.1 - 1 - 2 280 95
          FQ229429.1 - 1 - 2 280 95
          FQ229459.1 - 1 - 2 280 95
          FQ229557.1 - 1 - 2 280 95
          FQ229561.1 - 1 - 2 280 95
          FQ229574.1 - 1 - 2 280 95
          FQ229576.1 - 1 - 2 280 95
          FQ229614.1 - 1 - 2 280 95
          FQ229638.1 - 1 - 2 280 95
          FQ229640.1 - 1 - 2 280 95
          FQ229730.1 - 1 - 2 280 95
          FQ229752.1 - 1 - 2 282 95
          FQ229793.1 - 1 - 2 280 95
          FQ229848.1 - 2 - 3 245 95
          FQ229858.1 - 1 - 2 280 95
          FQ229873.1 - 1 - 2 280 95
          FQ229927.1 - 1 - 2 280 95
          FQ230015.1 - 1 - 2 280 95
          FQ230020.1 - 1 - 2 280 95
          FQ230030.1 - 1 - 2 280 95
          FQ230041.1 - 1 - 2 280 95
          FQ230049.1 - 1 - 2 280 95
          FQ230164.1 - 1 - 2 280 95
          FQ230220.1 - 1 - 2 280 95
          FQ230293.1 - 1 - 2 280 95
          FQ230398.1 - 1 - 2 280 95
          FQ230698.1 - 1 - 2 280 95
          FQ230986.1 - 1 - 2 281 95
          FQ231063.1 - 1 - 2 280 95
          FQ231580.1 - 2 - 3 209 95
          FQ231833.1 - 1 - 2 282 95
          FQ231996.1 - 1 - 2 280 95
          FQ232134.1 - 1 - 2 280 95
          FQ232888.1 - 1 - 2 280 95
          FQ233047.1 - 1 - 2 280 95
          FQ233467.1 - 1 - 2 266 95
          FQ233673.1 - 1 - 2 274 95
          FQ234189.1 - 1 - 2 280 95
          FQ234506.1 - 1 - 2 280 95
          U58829.1 - 1 - 2 243 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fth1

          Panther Classification:

          Molecular Function -

          storage protein

          Gene Ontology Categories:

          Function(s) Process(es)

          iron ion transport
          cellular iron ion homeostasis
          intracellular sequestering of iron ion
          immune response
          negative regulation of cell proliferation
          negative regulation of fibroblast proliferation
          oxidation-reduction process
          negative regulation of necrotic cell death
          ferroxidase activity
          protein binding
          ferric iron binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P075640 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119166 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13974 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13974 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P075640 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13974 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P292634 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P351546 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92537 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713827 - -
          Affymetrix Rat Gene 1.0 ST Array 10840606 - -
          Affymetrix Rat Gene 1.0 ST Array 10713829 - -
          Affymetrix Rat Gene 1.0 ST Array 10713826 - -
          Affymetrix Rat Gene 1.0 ST Array 10713830 - -
          Affymetrix Rat Gene 1.0 ST Array 10713828 - -
          Affymetrix Rat Gene 1.1 ST Array 10840605 - -
          Affymetrix Rat Gene 1.1 ST Array 10713825 - -
          Rat Genome 230 2.0 Array 1367565_a_at - -
          Rat Genome 230 2.0 Array 1367565_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367565_a_at - -
          Affymetrix HT Rat Focus Array 1367565_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100026236.rn.1 TC0100003179.rn.1 RTA TC
          Rat Expression Set 230 1367565_a_at - -
          Rat Expression Set 230 1367565_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713825 - -
          Affymetrix Rat Gene 1.0 ST Array 10840605 - -
          Rat Genome U34 Set rc_AI169802_at - -
          Rat Genome U34 Set rc_AA963209_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22084499 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359793 - -
          RatRef-12 Expression BeadChip ILMN_61561 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline