Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821330_g1
          See other AMY2A3 GE Assays ›
          Gene Symbol:
          Amy2a3
          Gene Name:
          amylase 2a3
          Gene Aliases:
          Amy2
          Chromosome Location:
          Chr.2: 216373674 - 216382260 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821330_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amy2a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031502.1 NP_113690.1 886 76
          GenBank mRNA FQ233367.1 - 5 - 6 881 76
          J00703.2 - 886 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Amy2a3

          Panther Classification:

          Molecular Function -

          amylase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          carbohydrate catabolic process
          alpha-amylase activity
          calcium ion binding
          protein binding
          chloride ion binding
          cation binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058472 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P057791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58551 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130295 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058472 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058472 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058472 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P057791 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P179767 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702203196 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66604 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29466 - -
          Agilent Rat cDNA Microarray (G4105A) 702203196 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826187 - -
          Affymetrix Rat Gene 1.0 ST Array 10826180 - -
          Affymetrix Rat Gene 1.0 ST Array 10826181 - -
          Affymetrix Rat Gene 1.0 ST Array 10826178 - -
          Affymetrix Rat Gene 1.0 ST Array 10826183 - -
          Affymetrix Rat Gene 1.0 ST Array 10826193 - -
          Affymetrix Rat Gene 1.0 ST Array 10826205 - -
          Affymetrix Rat Gene 1.0 ST Array 10826182 - -
          Affymetrix Rat Gene 1.0 ST Array 10826177 - -
          Affymetrix Rat Gene 1.0 ST Array 10826201 - -
          Affymetrix Rat Gene 1.0 ST Array 10826191 - -
          Affymetrix Rat Gene 1.0 ST Array 10826176 - -
          Affymetrix Rat Gene 1.0 ST Array 10826185 - -
          Affymetrix Rat Gene 1.0 ST Array 10826188 - -
          Affymetrix Rat Gene 1.0 ST Array 10826186 - -
          Affymetrix Rat Gene 1.0 ST Array 10826192 - -
          Affymetrix Rat Gene 1.0 ST Array 10826189 - -
          Affymetrix Rat Gene 1.1 ST Array 10826175 - -
          Affymetrix Rat Gene 1.1 ST Array 10826184 - -
          Rat Genome 230 2.0 Array 1369502_PM_a_at - -
          Rat Genome 230 2.0 Array 1369503_at - -
          Rat Genome 230 2.0 Array 1369503_PM_at - -
          Rat Genome 230 2.0 Array 1369502_a_at - -
          Affymetrix HT Rat Focus Array 1369503_at - -
          Affymetrix HT Rat Focus Array 1369502_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369502_a_at - -
          Affymetrix HT Rat Focus Array 1369503_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200035105.rn.1 TC0200005008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200035107.rn.1 TC0200005008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200035104.rn.1 TC0200005008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200035102.rn.1 TC0200005008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200035101.rn.1 TC0200005008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200035103.rn.1 TC0200005008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200035106.rn.1 TC0200005008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200035108.rn.1 TC0200005008.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200035109.rn.1 TC0200005008.rn.1 RTA TC
          Rat Expression Set 230 1369503_at - -
          Rat Expression Set 230 1369502_a_at - -
          Rat Expression Set 230 1369502_PM_a_at - -
          Rat Expression Set 230 1369503_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826175 - -
          Affymetrix Rat Gene 1.0 ST Array 10826184 - -
          Rat Genome U34 Set V01225mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21113435 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363412 - -
          RatRef-12 Expression BeadChip ILMN_48062 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline