Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821542_g1
          See other KCNK6 GE Assays ›
          Gene Symbol:
          Kcnk6 LOC100909725
          Gene Name:
          potassium channel, two pore domain subfamily K, member 6
          potassium channel subfamily K member 6-like
          Gene Aliases:
          Twik-2, Twik2
          -
          Chromosome Location:
          Chr.1: 88168438 - 88176610 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 87166455 - 87175348 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00821542_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnk6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053806.2 NP_446258.2 2 - 3 714 61
          GenBank mRNA AF281304.1 - 714 61
          AF281305.1 - 3 - 4 513 61
          Gene Symbol: LOC100909725
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003748812.3 XP_003748860.1 1241 61

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnk6
          Click here to find siRNAs for Gene LOC100909725

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure
          negative regulation of systemic arterial blood pressure
          regulation of resting membrane potential
          potassium ion transmembrane transport
          potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P738277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650228 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P738277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P738277 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P738277 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P738277 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421565 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61812 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68557 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720469 - -
          Affymetrix Rat Gene 1.0 ST Array 10720471 - -
          Affymetrix Rat Gene 1.0 ST Array 10720473 - -
          Affymetrix Rat Gene 1.0 ST Array 10720472 - -
          Affymetrix Rat Gene 1.0 ST Array 10720470 - -
          Affymetrix Rat Gene 1.1 ST Array 10720468 - -
          Rat Genome 230 2.0 Array 1387264_PM_at - -
          Rat Genome 230 2.0 Array 1370776_a_at - -
          Rat Genome 230 2.0 Array 1387264_at - -
          Rat Genome 230 2.0 Array 1370776_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370776_a_at - -
          Affymetrix HT Rat Focus Array 1370776_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387264_at - -
          Affymetrix HT Rat Focus Array 1387264_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100040605.rn.1 TC0100005098.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040607.rn.1 TC0100005098.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100040609.rn.1 TC0100005098.rn.1 RTA TC
          Rat Expression Set 230 1370776_a_at - -
          Rat Expression Set 230 1387264_at - -
          Rat Expression Set 230 1387264_PM_at - -
          Rat Expression Set 230 1370776_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10720468 - -
          Rat Genome U34 Set rc_AI177792_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22139336 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline