Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821667_g1
          See other DHFR GE Assays ›
          Gene Symbol:
          Dhfr
          Gene Name:
          dihydrofolate reductase
          Gene Aliases:
          Dhfr1
          Chromosome Location:
          Chr.2: 21931887 - 21958927 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821667_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dhfr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130400.2 NP_569084.1 5 - 6 567 105
          GenBank mRNA AF318150.1 - 5 - 6 568 105
          BC158583.1 - 494 9022774 105

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dhfr

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycine biosynthetic process
          one-carbon metabolic process
          nucleotide biosynthetic process
          response to methotrexate
          response to nicotine
          dihydrofolate metabolic process
          tetrahydrofolate metabolic process
          tetrahydrofolate biosynthetic process
          oxidation-reduction process
          mRNA binding
          dihydrofolate reductase activity
          drug binding
          NADP binding
          dihydrofolic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P247727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679560 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25606 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177539 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130965 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18447 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93294 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171336 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53230 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16737 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59204 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26460 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P247727 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P247727 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P247727 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P247727 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70227 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54027 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28605 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57384 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68747 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38299 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67865 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70956 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58550 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12999 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8819 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49850 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99250 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812457 - -
          Affymetrix Rat Gene 1.0 ST Array 10812460 - -
          Affymetrix Rat Gene 1.0 ST Array 10812462 - -
          Affymetrix Rat Gene 1.0 ST Array 10812461 - -
          Affymetrix Rat Gene 1.0 ST Array 10852650 - -
          Affymetrix Rat Gene 1.0 ST Array 10812463 - -
          Affymetrix Rat Gene 1.0 ST Array 10812459 - -
          Affymetrix Rat Gene 1.0 ST Array 10812458 - -
          Affymetrix Rat Gene 1.1 ST Array 10852649 - -
          Affymetrix Rat Gene 1.1 ST Array 10812456 - -
          Rat Genome 230 2.0 Array 1383536_PM_at - -
          Rat Genome 230 2.0 Array 1370075_at - -
          Rat Genome 230 2.0 Array 1370075_PM_at - -
          Rat Genome 230 2.0 Array 1383536_at - -
          Affymetrix HT Rat Focus Array 1370075_PM_at - -
          Affymetrix HT Rat Focus Array 1370075_at - -
          Affymetrix HT Rat Focus Array 1383536_PM_at - -
          Affymetrix HT Rat Focus Array 1383536_at - -
          Affymetrix Rat Clariom S Assay PSR0200000738.rn.1 TC0200000142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000737.rn.1 TC0200000142.rn.1 RTA TC
          Rat Expression Set 230 1370075_at - -
          Rat Expression Set 230 1370075_PM_at - -
          Rat Expression Set 230 1383536_at - -
          Rat Expression Set 230 1383536_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812456 - -
          Rat Genome U34 Set rc_AA900413_at - -
          Rat Genome U34 Set rc_AI232898_at - -
          Rat Genome U34 Set rc_AA900413_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22215106 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040525 - -
          RatRef-12 Expression BeadChip ILMN_59172 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline