Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821689_g1
          See other ATP6V0C GE Assays ›
          Gene Symbol:
          Atp6v0c
          Gene Name:
          ATPase, H+ transporting, lysosomal V0 subunit C
          Gene Aliases:
          Atp6c, Atp6l
          Chromosome Location:
          Chr.10: 13537031 - 13543407 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821689_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp6v0c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130823.3 NP_570836.1 1 - 2 234 119
          XM_006245898.1 XP_006245960.1 2 - 3 735 119
          GenBank mRNA BC063154.1 - 1 - 2 238 6918203 119
          D10874.1 - 1 - 2 175 119
          FQ211954.1 - 1 - 2 214 119
          FQ212145.1 - 1 - 2 239 119
          FQ212559.1 - 1 - 2 218 119
          FQ212733.1 - 1 - 2 218 119
          FQ213105.1 - 1 - 2 218 119
          FQ213455.1 - 1 - 2 218 119
          FQ214388.1 - 1 - 2 218 119
          FQ214503.1 - 1 - 2 239 119
          FQ216440.1 - 1 - 2 218 119
          FQ220211.1 - 1 - 2 218 119
          FQ220254.1 - 1 - 2 218 119
          FQ220713.1 - 1 - 2 239 119
          FQ220906.1 - 1 - 2 187 119
          FQ221569.1 - 1 - 2 229 119
          FQ229281.1 - 1 - 2 219 119
          FQ229453.1 - 1 - 2 214 119
          FQ229871.1 - 1 - 2 219 119
          FQ234225.1 - 1 - 2 239 119

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp6v0c

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          ATP hydrolysis coupled proton transport
          positive regulation of Wnt signaling pathway
          hydrogen ion transmembrane transporter activity
          ubiquitin protein ligase binding
          ATPase coupled ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P440280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835508 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88318 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P440280 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P440280 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P440280 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10171 - -
          Agilent Rat cDNA Microarray (G4105A) 700544301 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740997 - -
          Affymetrix Rat Gene 1.0 ST Array 10741000 - -
          Affymetrix Rat Gene 1.0 ST Array 10740998 - -
          Affymetrix Rat Gene 1.0 ST Array 10740999 - -
          Affymetrix Rat Gene 1.1 ST Array 10740996 - -
          Rat Genome 230 2.0 Array 1398755_PM_at - -
          Rat Genome 230 2.0 Array 1398755_at - -
          Affymetrix HT Rat Focus Array 1398755_at - -
          Affymetrix HT Rat Focus Array 1398755_PM_at - -
          Rat Expression Set 230 1398755_at - -
          Rat Expression Set 230 1398755_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10740996 - -
          Rat Genome U34 Set D10874_at - -
          Rat Genome U34 Set D10874_g_at - -
          Rat Genome U34 Set rc_AI177126_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21177319 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363169 - -
          RatRef-12 Expression BeadChip ILMN_53801 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline