Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821880_g1
          See other CITED1 GE Assays ›
          Gene Symbol:
          Cited1
          Gene Name:
          Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 1
          Gene Aliases:
          Msg1
          Chromosome Location:
          Chr.X: 72128997 - 72133692 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821880_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cited1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172055.2 NP_742052.2 2 - 3 227 77
          XM_006257097.2 XP_006257159.1 2 - 3 352 77
          XM_006257098.2 XP_006257160.1 3 - 4 916 77
          XM_006257099.2 XP_006257161.1 2 - 3 798 77
          XM_006257100.2 XP_006257162.1 2 - 3 242 77
          GenBank mRNA AF104399.1 - 1 - 2 85 77
          BC097276.1 - 2 - 3 227 7446633 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cited1

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          vasculogenesis
          branching involved in ureteric bud morphogenesis
          kidney development
          placenta development
          morphogenesis of an epithelium
          negative regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          regulation of transcription, DNA-templated
          nucleocytoplasmic transport
          transforming growth factor beta receptor signaling pathway
          brain development
          positive regulation of gene expression
          melanocyte differentiation
          response to lipopolysaccharide
          response to insulin
          response to cytokine
          response to interferon-gamma
          melanin biosynthetic process
          pigmentation
          negative regulation of neuron apoptotic process
          response to estrogen
          negative regulation of osteoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          response to cAMP
          SMAD protein signal transduction
          labyrinthine layer development
          spongiotrophoblast layer development
          response to interleukin-1
          response to interleukin-2
          response to interleukin-4
          response to interleukin-6
          response to interleukin-9
          response to interleukin-11
          response to parathyroid hormone
          response to transforming growth factor beta
          chromatin binding
          transcription coactivator activity
          protein C-terminus binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          LBD domain binding
          co-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P547771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24399 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P547771 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P547771 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P547771 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P547771 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700290209 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69425 - -
          Agilent Rat cDNA Microarray (G4105A) 700290209 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938732 - -
          Affymetrix Rat Gene 1.0 ST Array 10938731 - -
          Affymetrix Rat Gene 1.0 ST Array 10938733 - -
          Affymetrix Rat Gene 1.0 ST Array 10938730 - -
          Affymetrix Rat Gene 1.1 ST Array 10938729 - -
          Rat Genome 230 2.0 Array 1370805_at - -
          Rat Genome 230 2.0 Array 1370805_PM_at - -
          Affymetrix HT Rat Focus Array 1370805_at - -
          Affymetrix HT Rat Focus Array 1370805_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00012462.rn.1 TC0X00001961.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00012461.rn.1 TC0X00001961.rn.1 RTA TC
          Rat Expression Set 230 1370805_PM_at - -
          Rat Expression Set 230 1370805_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938729 - -
          Rat Genome U34 Set AF104399_at - -
          Rat Genome U34 Set AF104399_g_at - -
          Rat Genome U34 Set rc_AA964444_f_at - -
          Rat Genome U34 Set rc_AA964444_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21370507 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364035 - -
          RatRef-12 Expression BeadChip ILMN_55754 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline