Search
Search
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034911.1 | NP_001030083.1 | 5 - 6 | 752 | 60 | |
| GenBank mRNA | BC101866.1 | - | 5 - 6 | 752 | 7938969 | 60 |
| CK839597.1 | - | 2 - 3 | 60 | |||
Set Membership: |
|