Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01170103_g1
          See other RFC4 GE Assays ›
          Gene Symbol:
          Rfc4
          Gene Name:
          replication factor C (activator 1) 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 81358592 - 81373044 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01170103_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rfc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105869.2 NP_001099339.1 4 - 5 334 91
          GenBank mRNA BC168166.1 - 4 - 5 334 8374426 91
          FQ228232.1 - 4 - 5 366 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-directed DNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-dependent DNA replication
          biological_process
          molecular_function
          DNA clamp loader activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P132533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141480 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P132533 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P132533 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P132533 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P132533 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751979 - -
          Affymetrix Rat Gene 1.0 ST Array 10751974 - -
          Affymetrix Rat Gene 1.0 ST Array 10751976 - -
          Affymetrix Rat Gene 1.0 ST Array 10751981 - -
          Affymetrix Rat Gene 1.0 ST Array 10751975 - -
          Affymetrix Rat Gene 1.0 ST Array 10751980 - -
          Affymetrix Rat Gene 1.0 ST Array 10751978 - -
          Affymetrix Rat Gene 1.0 ST Array 10751977 - -
          Affymetrix Rat Gene 1.1 ST Array 10751973 - -
          Rat Genome 230 2.0 Array 1388458_at - -
          Rat Genome 230 2.0 Array 1388458_PM_at - -
          Affymetrix HT Rat Focus Array 1388458_PM_at - -
          Affymetrix HT Rat Focus Array 1388458_at - -
          Affymetrix Rat Clariom S Assay PSR1100004872.rn.1 TC1100000881.rn.1 RTA TC
          Rat Expression Set 230 1388458_PM_at - -
          Rat Expression Set 230 1388458_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751973 - -
          Rat Genome U34 Set rc_AI237756_at - -
          Rat Genome U34 Set rc_AA819500_at - -
          Rat Genome U34 Set rc_AA819500_g_at - -
          Rat Genome U34 Set rc_AI237756_at - -
          Rat Genome U34 Set rc_AA819500_at - -
          Rat Genome U34 Set rc_AA819500_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22205552 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354129 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline