Search
Search
| Gene Symbol: Il34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025766.1 | NP_001020937.1 | 2 - 3 | 535 | 83 | |
| XM_006255599.2 | XP_006255661.1 | 3 - 4 | 898 | 83 | ||
| XM_006255600.2 | XP_006255662.1 | 3 - 4 | 898 | 83 | ||
| XM_008772527.1 | XP_008770749.1 | 3 - 4 | 603 | 83 | ||
| GenBank mRNA | BC076391.1 | - | 3 - 4 | 661 | 83 | |
| BC098808.1 | - | 2 - 3 | 535 | 7376793 | 83 | |
Set Membership: |
|