Search
Search
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022507.1 | NP_071952.1 | 2 - 3 | 340 | 74 | |
| XM_008764337.1 | XP_008762559.1 | 2 - 3 | 241 | 74 | ||
| GenBank mRNA | J04532.1 | - | 2 - 3 | 340 | 74 | |
Set Membership: |
|