Search
Search
| Gene Symbol: Mtch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106488.2 | NP_001099958.2 | 754 | 114 | ||
| GenBank mRNA | BC158687.1 | - | 705 | 8375010 | 114 | |
| CK366546.1 | - | 151 | 114 | |||
Set Membership: |
|