Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01289321_g1
          See other MTFR1 GE Assays ›
          Gene Symbol:
          Mtfr1
          Gene Name:
          mitochondrial fission regulator 1
          Gene Aliases:
          RGD1304936
          Chromosome Location:
          Chr.2: 104005704 - 104040914 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01289321_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtfr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100977.1 NP_001094447.1 6 - 7 966 86
          XM_006232178.2 XP_006232240.1 7 - 8 1117 86
          XM_006232179.2 XP_006232241.1 7 - 8 1096 86
          XM_006232180.2 XP_006232242.1 6 - 7 1906 86
          GenBank mRNA EF221637.1 - 6 - 7 966 86
          FQ217237.1 - 7 - 8 1068 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          mitochondrial fission
          mitochondrion organization
          aerobic respiration

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P077865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29317 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P077865 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P077865 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P077865 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P396218 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1045157 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10814403 - -
          Affymetrix Rat Gene 1.0 ST Array 10814405 - -
          Affymetrix Rat Gene 1.0 ST Array 10839507 - -
          Affymetrix Rat Gene 1.0 ST Array 10814404 - -
          Affymetrix Rat Gene 1.0 ST Array 10814401 - -
          Affymetrix Rat Gene 1.0 ST Array 10814400 - -
          Affymetrix Rat Gene 1.0 ST Array 10814399 - -
          Affymetrix Rat Gene 1.0 ST Array 10814402 - -
          Affymetrix Rat Gene 1.1 ST Array 10839506 - -
          Affymetrix Rat Gene 1.1 ST Array 10814398 - -
          Rat Genome 230 2.0 Array 1393239_PM_at - -
          Rat Genome 230 2.0 Array 1393239_at - -
          Rat Genome 230 2.0 Array 1384046_PM_at - -
          Rat Genome 230 2.0 Array 1384046_at - -
          Affymetrix Rat Clariom S Assay PSR0200005119.rn.1 TC0200000844.rn.1 RTA TC
          Rat Expression Set 230 1393239_at - -
          Rat Expression Set 230 1393239_PM_at - -
          Rat Expression Set 230 1384046_at - -
          Rat Expression Set 230 1384046_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839506 - -
          Affymetrix Rat Gene 1.0 ST Array 10814398 - -
          Rat Genome U34 Set rc_AA892399_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21532111 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349571 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline