Search
Search
| Gene Symbol: Cab39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106924.1 | NP_001100394.1 | 2 - 3 | 531 | 117 | |
| XM_006245373.2 | XP_006245435.1 | 2 - 3 | 561 | 117 | ||
| XM_006245374.2 | XP_006245436.1 | 2 - 3 | 704 | 117 | ||
| XM_006245375.2 | XP_006245437.1 | 2 - 3 | 463 | 117 | ||
Set Membership: |
|