Search
Search
| Gene Symbol: Dnajb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109541.1 | NP_001103011.1 | 4 - 5 | 502 | 79 | |
| XM_006245276.1 | XP_006245338.1 | 5 - 6 | 561 | 79 | ||
| XM_006245278.2 | XP_006245340.1 | 5 - 6 | 583 | 79 | ||
| XM_006245279.2 | XP_006245341.1 | 5 - 6 | 510 | 79 | ||
| XM_006245280.1 | XP_006245342.1 | 5 - 6 | 561 | 79 | ||
| XM_006245281.2 | XP_006245343.1 | 4 - 5 | 510 | 79 | ||
| XM_006245283.1 | XP_006245345.1 | 5 - 6 | 561 | 79 | ||
| GenBank mRNA | BC166710.1 | - | 5 - 6 | 498 | 7930287 | 79 |
Set Membership: |
|