Search
Search
| Gene Symbol: Dnajb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109541.1 | NP_001103011.1 | 5 - 6 | 596 | 76 | |
| XM_006245276.1 | XP_006245338.1 | 6 - 7 | 655 | 76 | ||
| XM_006245278.2 | XP_006245340.1 | 6 - 7 | 677 | 76 | ||
| XM_006245279.2 | XP_006245341.1 | 6 - 7 | 604 | 76 | ||
| XM_006245280.1 | XP_006245342.1 | 6 - 7 | 655 | 76 | ||
| XM_006245281.2 | XP_006245343.1 | 5 - 6 | 604 | 76 | ||
| XM_006245283.1 | XP_006245345.1 | 6 - 7 | 655 | 76 | ||
| GenBank mRNA | BC166710.1 | - | 6 - 7 | 592 | 7930287 | 76 |
Set Membership: |
|