Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01400971_m1
          See other LOC100911733 GE Assays ›
          Gene Symbol:
          LOC100911733 Sp140
          Gene Name:
          nuclear body protein SP140-like
          SP140 nuclear body protein
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.9: 90930880 - 90938046 on Build Rnor_6.0
          Chromosome Location:
          Chr.9: 92618088 - 92665431 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01400971_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100911733
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006245337.2 XP_006245399.1 1 - 2 46 81
          Gene Symbol: Sp140
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012133.1 NP_001012133.1 2 - 3 658 81
          GenBank mRNA BC081971.1 - 2 - 3 658 7125855 81

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100911733
          Click here to find siRNAs for Gene Sp140

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          regulation of transcription, DNA-templated
          DNA binding
          chromatin binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225083 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116446 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128485 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104335 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85925 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015462 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015462 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015462 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62963 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81490 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28909 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924816 - -
          Affymetrix Rat Gene 1.0 ST Array 10924810 - -
          Affymetrix Rat Gene 1.0 ST Array 10924813 - -
          Affymetrix Rat Gene 1.0 ST Array 10924808 - -
          Affymetrix Rat Gene 1.0 ST Array 10924811 - -
          Affymetrix Rat Gene 1.0 ST Array 10924817 - -
          Affymetrix Rat Gene 1.0 ST Array 10924809 - -
          Affymetrix Rat Gene 1.0 ST Array 10924820 - -
          Affymetrix Rat Gene 1.0 ST Array 10924815 - -
          Affymetrix Rat Gene 1.0 ST Array 10924822 - -
          Affymetrix Rat Gene 1.0 ST Array 10924821 - -
          Affymetrix Rat Gene 1.0 ST Array 10924819 - -
          Affymetrix Rat Gene 1.0 ST Array 10924818 - -
          Affymetrix Rat Gene 1.0 ST Array 10924814 - -
          Affymetrix Rat Gene 1.0 ST Array 10924806 - -
          Affymetrix Rat Gene 1.0 ST Array 10924812 - -
          Affymetrix Rat Gene 1.0 ST Array 10924807 - -
          Affymetrix Rat Gene 1.0 ST Array 10924823 - -
          Affymetrix Rat Gene 1.1 ST Array 10924805 - -
          Rat Genome 230 2.0 Array 1374838_at - -
          Rat Genome 230 2.0 Array 1374838_PM_at - -
          Affymetrix HT Rat Focus Array 1374838_at - -
          Affymetrix HT Rat Focus Array 1374838_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900008562.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008563.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008576.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008583.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008584.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008578.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008577.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008580.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008579.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008582.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008575.rn.1 TC0900001015.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900008581.rn.1 TC0900001015.rn.1 RTA TC
          Rat Expression Set 230 1374838_at - -
          Rat Expression Set 230 1374838_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10924805 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21627108 - -
          ABI Rat Genome Survey Microarray 21649208 - -
          ABI Rat Genome Survey Microarray 21669329 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376263 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline