Search
Search

Cyp27a1| Gene Symbol: Cyp27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245175.1 | XP_006245237.1 | 5 - 6 | 904 | 94 | |
| GenBank mRNA | BC061848.1 | - | 5 - 6 | 1068 | 5621918 | 94 |
| M38566.1 | - | 5 - 6 | 1069 | 94 | ||
Set Membership: |
|