Search
Search
| Gene Symbol: Acadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012819.1 | NP_036951.1 | 9 - 10 | 1115 | 78 | |
| GenBank mRNA | BC062006.1 | - | 9 - 10 | 1113 | 5599828 | 78 |
| J05029.1 | - | 9 - 10 | 1126 | 78 | ||
| L11276.1 | - | 9 - 10 | 1115 | 78 | ||
Set Membership: |
|