Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01401633_m1
See other ABCA12 GE Assays ›
Gene Symbol:
Abca12
Gene Name:
ATP-binding cassette, subfamily A (ABC1), member 12
Gene Aliases:
-
Chromosome Location:
Chr.9: 78521576 - 78693403 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01401633_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Abca12
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_237242.9
XP_237242.6
33 - 34
5505
61
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Transporter
Recommended Assay
Rattus norvegicus
1700
Inventoried
Probe spans exons
Panther Classification:
Molecular Function -
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
biological_process
metabolic process
positive regulation of cholesterol efflux
keratinocyte differentiation
keratinization
secretion by cell
phospholipid efflux
ceramide transport
surfactant homeostasis
regulated exocytosis
lung alveolus development
lipid homeostasis
establishment of skin barrier
protein localization to plasma membrane
positive regulation of protein localization to cell surface
molecular_function
receptor binding
transporter activity
lipid transporter activity
ATP binding
ATPase activity
apolipoprotein A-I receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_44_P516160
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_44_P516160
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_44_P516160
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_44_P516160
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P516160
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Gene 1.1 ST Array
10928700
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
22148575
-
-
ABI Rat Genome Survey Microarray
22349549
-
-
Array Name
Probe IDs
Targets
Target Type
Illumina ratRef-12 v1.0 expression beadchip
ILMN_1350549
-
-
RatRef-12 Expression BeadChip
ILMN_47934
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit