Search
Search
| Gene Symbol: Duxbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252678.2 | XP_006252740.1 | 4 - 5 | 3345 | 62 | |
| XM_006252679.2 | XP_006252741.1 | 5 - 6 | 1346 | 62 | ||
| XM_008771030.1 | XP_008769252.1 | 2 - 3 | 837 | 62 | ||
| XM_008771031.1 | XP_008769253.1 | 3 - 4 | 4020 | 62 | ||
| XM_224643.6 | XP_224643.1 | 4 - 5 | 2021 | 62 | ||
Set Membership: |
|