Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01402377_m1
          See other ERCC3 GE Assays ›
          Gene Symbol:
          Ercc3
          Gene Name:
          excision repair cross-complementation group 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 25037668 - 25068380 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01402377_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ercc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001031644.1 NP_001026814.1 14 - 15 2259 64
          GenBank mRNA BC098856.1 - 14 - 15 2259 7379944 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000232.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ercc3

          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA helicase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide-excision repair, DNA duplex unwinding
          response to hypoxia
          DNA topological change
          DNA repair
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair
          transcription from RNA polymerase II promoter
          protein phosphorylation
          apoptotic process
          response to oxidative stress
          protein localization
          response to UV
          UV protection
          nucleotide-excision repair, DNA incision
          hair cell differentiation
          positive regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          regulation of mitotic cell cycle phase transition
          DNA binding
          ATP-dependent DNA helicase activity
          protein kinase activity
          protein binding
          ATP binding
          GTP binding
          protein C-terminus binding
          DNA-dependent ATPase activity
          transcription factor binding
          RNA polymerase II carboxy-terminal domain kinase activity
          ATPase activity
          dATP binding
          peptide binding
          3'-5' DNA helicase activity
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P127649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170456 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56946 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127649 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127649 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P127649 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P440905 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31130 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61864 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74193 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10800757 - -
          Affymetrix Rat Gene 1.0 ST Array 10800753 - -
          Affymetrix Rat Gene 1.0 ST Array 10800749 - -
          Affymetrix Rat Gene 1.0 ST Array 10800746 - -
          Affymetrix Rat Gene 1.0 ST Array 10800750 - -
          Affymetrix Rat Gene 1.0 ST Array 10800754 - -
          Affymetrix Rat Gene 1.0 ST Array 10800751 - -
          Affymetrix Rat Gene 1.0 ST Array 10800758 - -
          Affymetrix Rat Gene 1.0 ST Array 10800756 - -
          Affymetrix Rat Gene 1.0 ST Array 10800752 - -
          Affymetrix Rat Gene 1.0 ST Array 10800755 - -
          Affymetrix Rat Gene 1.0 ST Array 10800745 - -
          Affymetrix Rat Gene 1.0 ST Array 10800747 - -
          Affymetrix Rat Gene 1.0 ST Array 10800748 - -
          Affymetrix Rat Gene 1.0 ST Array 10800759 - -
          Affymetrix Rat Gene 1.1 ST Array 10800744 - -
          Rat Genome 230 2.0 Array 1374011_at - -
          Rat Genome 230 2.0 Array 1374011_PM_at - -
          Affymetrix HT Rat Focus Array 1374011_at - -
          Affymetrix HT Rat Focus Array 1374011_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800001616.rn.1 TC1800000232.rn.1 RTA TC
          Rat Expression Set 230 1374011_PM_at - -
          Rat Expression Set 230 1374011_at - -
          Affymetrix Rat Gene 1.0 ST Array 10800744 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22135275 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373561 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline