Search
Search
| Gene Symbol: C9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057146.1 | NP_476487.1 | 5 - 6 | 658 | 108 | |
| GenBank mRNA | BC091127.1 | - | 5 - 6 | 654 | 7324872 | 108 |
| U49071.1 | - | 5 - 6 | 640 | 108 | ||
| U52948.1 | - | 5 - 6 | 658 | 108 | ||
Set Membership: |
|