Search
Search
| Gene Symbol: Mon1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126284.1 | NP_001119756.1 | 4 - 5 | 1552 | 86 | |
| XM_006243782.1 | XP_006243844.1 | 4 - 5 | 1441 | 86 | ||
| XM_006243783.2 | XP_006243845.1 | 4 - 5 | 1686 | 86 | ||
| XM_008766536.1 | XP_008764758.1 | 3 - 4 | 1201 | 86 | ||
| GenBank mRNA | BC161958.1 | - | 4 - 5 | 1571 | 9027405 | 86 |
Set Membership: |
|