Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108772.1 | NP_001102242.1 | 6 - 7 | 901 | 64 | |
XM_006243482.2 | XP_006243544.1 | 6 - 7 | 913 | 64 | ||
XM_008766451.1 | XP_008764673.1 | 6 - 7 | 929 | 64 | ||
GenBank mRNA | BC166838.1 | - | 6 - 7 | 823 | 9027278 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |