Search
Search
| Gene Symbol: Bcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107084.1 | NP_001100554.1 | 7 - 8 | 1896 | 69 | |
| XM_008768799.1 | XP_008767021.1 | 6 - 7 | 2769 | 69 | ||
| GenBank mRNA | BC166425.1 | - | 7 - 8 | 1796 | 7097735 | 69 |
Set Membership: |
|