Search
Search

Serpind1| Gene Symbol: Serpind1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024382.1 | NP_077358.1 | 2 - 3 | 104 | 81 | |
| GenBank mRNA | BC091145.1 | - | 2 - 3 | 84 | 7372906 | 81 |
| X74549.1 | - | 2 - 3 | 104 | 81 | ||
Set Membership: |
|