Search
Search
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012727.3 | NP_036859.2 | 4 - 5 | 459 | 98 | |
| XM_006254523.2 | XP_006254585.1 | 3 - 4 | 401 | 98 | ||
| GenBank mRNA | BC128706.1 | - | 4 - 5 | 459 | 7455982 | 98 |
| M63334.1 | - | 353 | 98 | |||
| M64757.1 | - | 2 - 3 | 449 | 98 | ||
| S65840.1 | - | 3 - 4 | 384 | 98 | ||
Set Membership: |
|