Search
Search

Arhgap26| Gene Symbol: Arhgap26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107389.1 | NP_001100859.1 | 16 - 17 | 1751 | 94 | |
| XM_006254660.2 | XP_006254722.1 | 16 - 17 | 1820 | 94 | ||
| XM_006254662.2 | XP_006254724.1 | 16 - 17 | 1816 | 94 | ||
| XM_006254663.2 | XP_006254725.1 | 16 - 17 | 1815 | 94 | ||
| XM_008772068.1 | XP_008770290.1 | 19 - 20 | 2049 | 94 | ||
Set Membership: |
|