Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01407182_m1
          See other RIPPLY3 GE Assays ›
          Gene Symbol:
          Ripply3
          Gene Name:
          ripply transcriptional repressor 3
          Gene Aliases:
          Dscr6
          Chromosome Location:
          Chr.11: 34557679 - 34565776 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01407182_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ripply3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105892.1 NP_001099362.1 3 - 4 314 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000320.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          negative regulation of transcription from RNA polymerase II promoter
          heart development
          negative regulation of cell proliferation
          pharyngeal system development

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P380384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126670 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55097 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P380384 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P380384 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P380384 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P380384 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750422 - -
          Affymetrix Rat Gene 1.0 ST Array 10750425 - -
          Affymetrix Rat Gene 1.0 ST Array 10750424 - -
          Affymetrix Rat Gene 1.0 ST Array 10750423 - -
          Affymetrix Rat Gene 1.1 ST Array 10750421 - -
          Rat Genome 230 2.0 Array 1391648_at - -
          Rat Genome 230 2.0 Array 1391648_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100001490.rn.1 TC1100000320.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100001491.rn.1 TC1100000320.rn.1 RTA TC
          Rat Expression Set 230 1391648_PM_at - -
          Rat Expression Set 230 1391648_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750421 - -
          Rat Genome U34 Set rc_AA944415_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20790717 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359522 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline