Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01408148_g1
          See other OAZ1 GE Assays ›
          Gene Symbol:
          Oaz1
          Gene Name:
          ornithine decarboxylase antizyme 1
          Gene Aliases:
          ODC-Az, ODCAC, Oaz
          Chromosome Location:
          Chr.7: 11752127 - 11754587 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01408148_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oaz1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001297557.1 NP_001284486.1 2 - 3 370 93
          NM_001301048.1 NP_001287977.1 2 - 3 364 93
          NM_139081.2 NP_620781.1 2 - 3 370 93
          GenBank mRNA D10706.1 - 2 - 3 342 93
          FQ211375.1 - 2 - 3 370 93
          FQ213794.1 - 2 - 3 370 93
          FQ214019.1 - 2 - 3 365 93
          FQ214051.1 - 2 - 3 365 93
          FQ214373.1 - 2 - 3 365 93
          FQ214458.1 - 2 - 3 365 93
          FQ216817.1 - 2 - 3 370 93
          FQ218486.1 - 2 - 3 365 93
          FQ218990.1 - 2 - 3 365 93
          FQ220491.1 - 2 - 3 365 93
          FQ220580.1 - 2 - 3 370 93
          FQ225116.1 - 2 - 3 365 93
          FQ225909.1 - 2 - 3 365 93
          FQ225949.1 - 2 - 3 368 93
          FQ229954.1 - 2 - 3 365 93
          FQ232667.1 - 2 - 3 365 93
          FQ232997.1 - 2 - 3 365 93
          FQ233723.1 - 2 - 3 365 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oaz1

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          polyamine metabolic process
          polyamine biosynthetic process
          transport
          negative regulation of catalytic activity
          positive regulation of protein catabolic process
          negative regulation of protein homodimerization activity
          positive regulation of intracellular protein transport
          negative regulation of polyamine transmembrane transport
          ornithine decarboxylase inhibitor activity
          enzyme binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152265 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058221 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058221 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058221 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P483419 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700517760 - -
          Agilent Rat cDNA Microarray (G4105A) 700517760 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900401 - -
          Affymetrix Rat Gene 1.0 ST Array 10900404 - -
          Affymetrix Rat Gene 1.0 ST Array 10753459 - -
          Affymetrix Rat Gene 1.0 ST Array 10900402 - -
          Affymetrix Rat Gene 1.0 ST Array 10900400 - -
          Affymetrix Rat Gene 1.0 ST Array 10900403 - -
          Affymetrix Rat Gene 1.1 ST Array 10753458 - -
          Affymetrix Rat Gene 1.1 ST Array 10900399 - -
          Rat Genome 230 2.0 Array 1372241_PM_at - -
          Rat Genome 230 2.0 Array 1372241_at - -
          Affymetrix HT Rat Focus Array 1372241_PM_at - -
          Affymetrix HT Rat Focus Array 1372241_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900399 - -
          Affymetrix Rat Gene 1.0 ST Array 10753458 - -
          Rat Genome U34 Set D10706_s_at - -
          Rat Genome U34 Set D10706cds#2_s_at - -
          Rat Genome U34 Set D10706cds#3_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21394797 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349040 - -
          RatRef-12 Expression BeadChip ILMN_65298 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline