Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01408150_g1
          See other OAZ1 GE Assays ›
          Gene Symbol:
          Oaz1
          Gene Name:
          ornithine decarboxylase antizyme 1
          Gene Aliases:
          ODC-Az, ODCAC, Oaz
          Chromosome Location:
          Chr.7: 11752127 - 11754587 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01408150_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oaz1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001297557.1 NP_001284486.1 4 - 5 629 105
          NM_001301048.1 NP_001287977.1 4 - 5 623 105
          NM_139081.2 NP_620781.1 4 - 5 629 105
          GenBank mRNA D10706.1 - 4 - 5 601 105
          FQ211375.1 - 4 - 5 629 105
          FQ213504.1 - 4 - 5 629 105
          FQ213794.1 - 4 - 5 629 105
          FQ214019.1 - 4 - 5 624 105
          FQ214051.1 - 4 - 5 624 105
          FQ216817.1 - 4 - 5 629 105
          FQ218486.1 - 4 - 5 624 105
          FQ218990.1 - 4 - 5 624 105
          FQ220491.1 - 4 - 5 624 105
          FQ220580.1 - 4 - 5 629 105
          FQ225116.1 - 4 - 5 624 105
          FQ225909.1 - 4 - 5 624 105
          FQ225949.1 - 4 - 5 627 105
          FQ229954.1 - 4 - 5 624 105
          FQ230452.1 - 4 - 5 624 105
          FQ231088.1 - 4 - 5 624 105
          FQ232667.1 - 4 - 5 624 105
          FQ232997.1 - 4 - 5 624 105
          FQ233723.1 - 4 - 5 624 105

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oaz1

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          polyamine metabolic process
          polyamine biosynthetic process
          transport
          negative regulation of catalytic activity
          positive regulation of protein catabolic process
          negative regulation of protein homodimerization activity
          positive regulation of intracellular protein transport
          negative regulation of polyamine transmembrane transport
          ornithine decarboxylase inhibitor activity
          enzyme binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152265 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058221 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058221 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058221 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P483419 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700517760 - -
          Agilent Rat cDNA Microarray (G4105A) 700517760 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900404 - -
          Affymetrix Rat Gene 1.0 ST Array 10900400 - -
          Affymetrix Rat Gene 1.0 ST Array 10900402 - -
          Affymetrix Rat Gene 1.0 ST Array 10753459 - -
          Affymetrix Rat Gene 1.0 ST Array 10900403 - -
          Affymetrix Rat Gene 1.0 ST Array 10900401 - -
          Affymetrix Rat Gene 1.1 ST Array 10900399 - -
          Affymetrix Rat Gene 1.1 ST Array 10753458 - -
          Rat Genome 230 2.0 Array 1372241_at - -
          Rat Genome 230 2.0 Array 1372241_PM_at - -
          Affymetrix HT Rat Focus Array 1372241_at - -
          Affymetrix HT Rat Focus Array 1372241_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700016794.rn.1 TC0700002134.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016798.rn.1 TC0700002134.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016796.rn.1 TC0700002134.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016797.rn.1 TC0700002134.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016795.rn.1 TC0700002134.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10900399 - -
          Affymetrix Rat Gene 1.0 ST Array 10753458 - -
          Rat Genome U34 Set D10706cds#2_s_at - -
          Rat Genome U34 Set D10706cds#3_s_at - -
          Rat Genome U34 Set D10706_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21394797 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349040 - -
          RatRef-12 Expression BeadChip ILMN_65298 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline